Assay Details
| Gene Symbol: ubl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017765.1 | NP_001017765.1 | 4 - 5 | 574 | 96 | |
| GenBank mRNA | BC055150.1 | - | 4 - 5 | 615 | 96 | |
| BC093190.1 | - | 4 - 5 | 574 | 96 | ||
| BC164699.1 | - | 4 - 5 | 506 | 96 | ||
More Information
Set Membership: |
|

ubl7a
Genomic Map