Assay Details
| Gene Symbol: ubl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017765.1 | NP_001017765.1 | 6 - 7 | 770 | 59 | |
| GenBank mRNA | BC055150.1 | - | 6 - 7 | 811 | 59 | |
| BC093190.1 | - | 6 - 7 | 770 | 59 | ||
| BC164699.1 | - | 6 - 7 | 702 | 59 | ||
More Information
Set Membership: |
|

ubl7a
Genomic Map