Assay Details
| Gene Symbol: ubl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017765.1 | NP_001017765.1 | 7 - 8 | 817 | 72 | |
| GenBank mRNA | BC093190.1 | - | 7 - 8 | 817 | 72 | |
| BC164699.1 | - | 7 - 8 | 749 | 72 | ||
More Information
Set Membership: |
|

ubl7a
Genomic Map