Assay Details
Gene Symbol: trhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044346.1 | NP_001037811.1 | 1 - 2 | 387 | 66 | |
XM_005159096.4 | XP_005159153.1 | 2 - 3 | 843 | 66 | ||
GenBank mRNA | BC171609.1 | - | 1 - 2 | 460 | 66 | |
BC171611.1 | - | 1 - 2 | 460 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
