Assay Details
Gene Symbol: trhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044346.1 | NP_001037811.1 | 2 - 3 | 803 | 61 | |
XM_005159096.4 | XP_005159153.1 | 3 - 4 | 1259 | 61 | ||
GenBank mRNA | BC171609.1 | - | 2 - 3 | 876 | 61 | |
BC171611.1 | - | 2 - 3 | 876 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
