Assay Details
Gene Symbol: notch1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131441.1 | NP_571516.1 | 21 - 22 | 3792 | 122 | |
GenBank mRNA | X69088.1 | - | 3792 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
