Assay Details
| Gene Symbol: notch1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131441.1 | NP_571516.1 | 14 - 15 | 2620 | 83 | |
| GenBank mRNA | X69088.1 | - | 2620 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

notch1a| Gene Symbol: notch1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131441.1 | NP_571516.1 | 14 - 15 | 2620 | 83 | |
| GenBank mRNA | X69088.1 | - | 2620 | 83 | ||
Set Membership: |
|