Assay Details
| Gene Symbol: star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131663.1 | NP_571738.1 | 5 - 6 | 692 | 60 | |
| GenBank mRNA | AF220435.1 | - | 5 - 6 | 692 | 60 | |
| BC075967.1 | - | 5 - 6 | 684 | 60 | ||
More Information
Set Membership: |
|

star
Genomic Map