Assay Details
| Gene Symbol: rrp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199762.1 | NP_956056.1 | 4 - 5 | 625 | 59 | |
| GenBank mRNA | BC046037.1 | - | 4 - 5 | 625 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rrp15| Gene Symbol: rrp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199762.1 | NP_956056.1 | 4 - 5 | 625 | 59 | |
| GenBank mRNA | BC046037.1 | - | 4 - 5 | 625 | 59 | |
Set Membership: |
|