Assay Details
| Gene Symbol: lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201070.3 | NP_957364.2 | 2 - 3 | 395 | 65 | |
| XM_009296529.3 | XP_009294804.1 | 2 - 3 | 406 | 65 | ||
| GenBank mRNA | BC049043.1 | - | 3 - 4 | 283 | 65 | |
| EB790305.1 | - | 2 - 3 | 184 | 65 | ||
| EH445290.1 | - | 2 - 3 | 185 | 65 | ||
More Information
Set Membership: |
|

lmcd1
Genomic Map