Assay Details
| Gene Symbol: per1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212439.2 | NP_997604.2 | 14 - 15 | 3181 | 66 | |
| GenBank mRNA | AY597250.1 | - | 14 - 15 | 3181 | 66 | |
| BC163718.1 | - | 13 - 14 | 66 | |||
More Information
Set Membership: |
|

per1b
Genomic Map