Assay Details
| Gene Symbol: chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213320.1 | NP_998485.1 | 5 - 6 | 561 | 69 | |
| GenBank mRNA | BC066696.1 | - | 5 - 6 | 561 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chmp3| Gene Symbol: chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213320.1 | NP_998485.1 | 5 - 6 | 561 | 69 | |
| GenBank mRNA | BC066696.1 | - | 5 - 6 | 561 | 69 | |
Set Membership: |
|