Assay Details
Gene Symbol: rfc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214737.2 | NP_999902.2 | 6 - 7 | 659 | 82 | |
GenBank mRNA | AY648805.1 | - | 6 - 7 | 659 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
