Assay Details
| Gene Symbol: mettl21e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002361.2 | NP_001002361.2 | 4 - 5 | 543 | 69 | |
| GenBank mRNA | BC076050.1 | - | 3 - 4 | 506 | 69 | |
| EB850753.1 | - | 4 - 5 | 534 | 69 | ||
More Information
Set Membership: |
|

mettl21e
Genomic Map