Assay Details
| Gene Symbol: cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003736.1 | NP_001003736.1 | 1 | 345 | 99 | |
| GenBank mRNA | BC077160.1 | - | 1 | 345 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyp8b1| Gene Symbol: cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003736.1 | NP_001003736.1 | 1 | 345 | 99 | |
| GenBank mRNA | BC077160.1 | - | 1 | 345 | 99 | |
Set Membership: |
|