Assay Details
| Gene Symbol: fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017876.1 | NP_001017876.1 | 1514 | 69 | ||
| GenBank mRNA | BC093429.1 | - | 1514 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fsip1| Gene Symbol: fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017876.1 | NP_001017876.1 | 1514 | 69 | ||
| GenBank mRNA | BC093429.1 | - | 1514 | 69 | ||
Set Membership: |
|