Assay Details
| Gene Symbol: snapin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045148.1 | NP_001038613.1 | 2 - 3 | 161 | 84 | |
| GenBank mRNA | BC139575.1 | - | 2 - 3 | 189 | 84 | |
More Information
Set Membership: |
|

snapin
Genomic Map