Assay Details
| Gene Symbol: edn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131519.1 | NP_571594.1 | 3 - 4 | 422 | 127 | |
| GenBank mRNA | AF281858.1 | - | 3 - 4 | 422 | 127 | |
| BC162539.1 | - | 3 - 4 | 376 | 127 | ||
More Information
Set Membership: |
|

edn1
Genomic Map