Assay Details
| Gene Symbol: tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200281.1 | NP_956575.1 | 5 - 6 | 614 | 62 | |
| XM_021469763.1 | XP_021325438.1 | 5 - 6 | 564 | 62 | ||
| GenBank mRNA | AY398411.1 | - | 5 - 6 | 651 | 62 | |
| BC049345.1 | - | 5 - 6 | 614 | 62 | ||
More Information
Set Membership: |
|

tspan31
Genomic Map