Assay Details
Gene Symbol: abcf2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201315.2 | NP_958472.2 | 12 - 13 | 1547 | 69 | |
GenBank mRNA | BC047181.1 | - | 12 - 13 | 1522 | 69 | |
BC066505.1 | - | 12 - 13 | 1451 | 69 | ||
BC165771.1 | - | 11 - 12 | 1426 | 69 | ||
BI709967.1 | - | 4 - 5 | 282 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
