Assay Details
| Gene Symbol: abcf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201315.2 | NP_958472.2 | 14 - 15 | 1880 | 85 | |
| GenBank mRNA | BC047181.1 | - | 14 - 15 | 85 | ||
| BC066505.1 | - | 14 - 15 | 1784 | 85 | ||
| BC165771.1 | - | 13 - 14 | 85 | |||
| BE557347.1 | - | 1 - 2 | 145 | 85 | ||
More Information
Set Membership: |
|

abcf2a
Genomic Map