Assay Details
| Gene Symbol: abcf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201315.2 | NP_958472.2 | 2 - 3 | 306 | 97 | |
| GenBank mRNA | BC047181.1 | - | 2 - 3 | 281 | 97 | |
| BC066505.1 | - | 2 - 3 | 210 | 97 | ||
| BC165771.1 | - | 1 - 2 | 185 | 97 | ||
| EH556650.1 | - | 2 - 3 | 345 | 97 | ||
More Information
Set Membership: |
|

abcf2a
Genomic Map