Assay Details
| Gene Symbol: arcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201459.1 | NP_958867.1 | 2 - 3 | 438 | 99 | |
| GenBank mRNA | BC045318.1 | - | 2 - 3 | 438 | 99 | |
| BC165628.1 | - | 1 - 2 | 303 | 99 | ||
More Information
Set Membership: |
|

arcn1a
Genomic Map