Assay Details
| Gene Symbol: arcn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201459.1 | NP_958867.1 | 7 - 8 | 1287 | 76 | |
| GenBank mRNA | BC045318.1 | - | 7 - 8 | 1287 | 76 | |
| BC165628.1 | - | 6 - 7 | 1152 | 76 | ||
More Information
Set Membership: |
|

arcn1a
Genomic Map