Assay Details
| Gene Symbol: tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213298.2 | NP_998463.1 | 2 - 3 | 673 | 92 | |
| GenBank mRNA | BC065312.1 | - | 2 - 3 | 618 | 92 | |
| EH541175.1 | - | 2 - 3 | 706 | 92 | ||
More Information
Set Membership: |
|

tmub1
Genomic Map