Assay Details
| Gene Symbol: mrpl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004683.1 | NP_001004683.1 | 2 - 3 | 457 | 125 | |
| GenBank mRNA | BC081486.1 | - | 2 - 3 | 457 | 125 | |
More Information
Set Membership: |
|

mrpl17
Genomic Map