Assay Details
| Gene Symbol: rrp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017579.1 | NP_001017579.1 | 2 - 3 | 204 | 79 | |
| GenBank mRNA | BC092724.1 | - | 2 - 3 | 204 | 79 | |
More Information
Set Membership: |
|

rrp7a
Genomic Map