Assay Details
| Gene Symbol: htr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044743.1 | NP_001038208.1 | 1 - 2 | 297 | 74 | |
| GenBank mRNA | BC162678.1 | - | 1 - 2 | 284 | 74 | |
| BC162703.1 | - | 1 - 2 | 284 | 74 | ||
| DQ864496.1 | - | 1 - 2 | 297 | 74 | ||
More Information
Set Membership: |
|

htr2b
Genomic Map