Assay Details
| Gene Symbol: wnt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079834.1 | NP_001073303.1 | 3 - 4 | 568 | 59 | |
| GenBank mRNA | BC150218.1 | - | 3 - 4 | 847 | 59 | |
| BC163085.1 | - | 3 - 4 | 563 | 59 | ||
| BC163092.1 | - | 3 - 4 | 563 | 59 | ||
| DQ465921.1 | - | 3 - 4 | 568 | 59 | ||
More Information
Set Membership: |
|

wnt5a
Genomic Map