Assay Details
| Gene Symbol: tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080026.1 | NP_001073495.1 | 3 - 4 | 751 | 120 | |
| XM_005166193.4 | XP_005166250.1 | 3 - 4 | 713 | 120 | ||
| XM_017356508.2 | XP_017211997.1 | 3 - 4 | 1291 | 120 | ||
| GenBank mRNA | BC122136.1 | - | 3 - 4 | 751 | 120 | |
More Information
Set Membership: |
|

tefm
Genomic Map