Assay Details
| Gene Symbol: glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152965.1 | NP_694497.1 | 6 - 7 | 928 | 64 | |
| XM_021477569.1 | XP_021333244.1 | 6 - 7 | 1077 | 64 | ||
| GenBank mRNA | AJ308516.1 | - | 6 - 7 | 928 | 64 | |
More Information
Set Membership: |
|

glra3
Genomic Map