Assay Details
| Gene Symbol: ccz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200134.1 | NP_956428.1 | 14 - 15 | 1231 | 107 | |
| XM_017358438.2 | XP_017213927.1 | 14 - 15 | 1182 | 107 | ||
| GenBank mRNA | BC044374.1 | - | 14 - 15 | 1231 | 107 | |
| BC152682.1 | - | 14 - 15 | 1276 | 107 | ||
More Information
Set Membership: |
|

ccz1
Genomic Map