Assay Details
| Gene Symbol: c8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200863.1 | NP_957157.1 | 6 - 7 | 692 | 84 | |
| GenBank mRNA | BC062865.1 | - | 6 - 7 | 692 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

c8g| Gene Symbol: c8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200863.1 | NP_957157.1 | 6 - 7 | 692 | 84 | |
| GenBank mRNA | BC062865.1 | - | 6 - 7 | 692 | 84 | |
Set Membership: |
|