Assay Details
| Gene Symbol: mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213369.1 | NP_998534.1 | 3 - 4 | 394 | 86 | |
| GenBank mRNA | BC046085.1 | - | 3 - 4 | 394 | 86 | |
More Information
Set Membership: |
|

mospd1
Genomic Map