Assay Details
Gene Symbol: hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328494.1 | NP_001315423.1 | 1 - 2 | 225 | 70 | |
NR_137312.1 | 1 - 2 | 225 | 70 | |||
XM_005155542.3 | XP_005155599.1 | 1 - 2 | 231 | 70 | ||
XM_021479148.1 | XP_021334823.1 | 1 - 2 | 230 | 70 | ||
XM_021479149.1 | XP_021334824.1 | 1 - 2 | 230 | 70 | ||
XM_021479150.1 | XP_021334825.1 | 1 - 2 | 231 | 70 | ||
GenBank mRNA | BC055525.1 | - | 1 - 2 | 225 | 70 | |
EH541511.1 | - | 1 - 2 | 157 | 70 | ||
EH541767.1 | - | 1 - 2 | 203 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
