Assay Details
| Gene Symbol: desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003532.1 | NP_001003532.1 | 4 - 5 | 832 | 99 | |
| GenBank mRNA | BC078248.1 | - | 4 - 5 | 832 | 99 | |
More Information
Set Membership: |
|

desi2
Genomic Map