Assay Details
Gene Symbol: picalmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003741.1 | NP_001003741.1 | 10 - 11 | 1353 | 67 | |
XM_009291635.3 | XP_009289910.1 | 11 - 12 | 1510 | 67 | ||
XM_009291636.3 | XP_009289911.1 | 10 - 11 | 1498 | 67 | ||
XM_009291637.3 | XP_009289912.1 | 11 - 12 | 1507 | 67 | ||
XM_009291642.3 | XP_009289917.1 | 10 - 11 | 1491 | 67 | ||
XM_009291643.3 | XP_009289918.1 | 10 - 11 | 1494 | 67 | ||
GenBank mRNA | BC079523.1 | - | 10 - 11 | 1353 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
