Assay Details
Gene Symbol: picalmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003741.1 | NP_001003741.1 | 9 - 10 | 1233 | 92 | |
XM_009291635.3 | XP_009289910.1 | 10 - 11 | 1390 | 92 | ||
XM_009291636.3 | XP_009289911.1 | 9 - 10 | 1378 | 92 | ||
XM_009291637.3 | XP_009289912.1 | 10 - 11 | 1387 | 92 | ||
XM_009291638.3 | XP_009289913.1 | 10 - 11 | 1392 | 92 | ||
XM_009291639.3 | XP_009289914.1 | 10 - 11 | 1390 | 92 | ||
XM_009291640.3 | XP_009289915.1 | 10 - 11 | 1388 | 92 | ||
XM_009291641.3 | XP_009289916.1 | 10 - 11 | 1387 | 92 | ||
XM_009291642.3 | XP_009289917.1 | 9 - 10 | 1371 | 92 | ||
XM_009291643.3 | XP_009289918.1 | 9 - 10 | 1374 | 92 | ||
XM_017359233.2 | XP_017214722.1 | 10 - 11 | 1393 | 92 | ||
XM_017359234.2 | XP_017214723.1 | 9 - 10 | 1380 | 92 | ||
XM_017359235.2 | XP_017214724.1 | 10 - 11 | 1392 | 92 | ||
XM_017359236.2 | XP_017214725.1 | 10 - 11 | 1390 | 92 | ||
GenBank mRNA | BC079523.1 | - | 9 - 10 | 1233 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
