Assay Details
| Gene Symbol: bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005575.1 | NP_001005575.1 | 4 - 5 | 851 | 75 | |
| XM_005159238.4 | XP_005159295.1 | 4 - 5 | 955 | 75 | ||
| XM_005159239.4 | XP_005159296.1 | 4 - 5 | 953 | 75 | ||
| GenBank mRNA | AY296057.1 | - | 4 - 5 | 851 | 75 | |
| BC163544.1 | - | 4 - 5 | 831 | 75 | ||
More Information
Set Membership: |
|

bcl9l
Genomic Map