Assay Details
| Gene Symbol: crata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005587.1 | NP_001005587.1 | 13 - 14 | 1709 | 101 | |
| GenBank mRNA | BC081647.1 | - | 13 - 14 | 1709 | 101 | |
More Information
Set Membership: |
|

crata
Genomic Map