Assay Details
| Gene Symbol: crata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005587.1 | NP_001005587.1 | 6 - 7 | 852 | 110 | |
| GenBank mRNA | BC081647.1 | - | 6 - 7 | 852 | 110 | |
More Information
Set Membership: |
|

crata
Genomic Map