Assay Details
| Gene Symbol: dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007061.1 | NP_001007062.1 | 5 - 6 | 469 | 61 | |
| GenBank mRNA | BC083210.1 | - | 5 - 6 | 469 | 61 | |
| BC165079.1 | - | 5 - 6 | 490 | 61 | ||
More Information
Set Membership: |
|

dolpp1
Genomic Map