Assay Details
| Gene Symbol: fgf12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077782.1 | NP_001071250.1 | 1 - 2 | 883 | 63 | |
| GenBank mRNA | AB195618.2 | - | 1 - 2 | 194 | 63 | |
| BC124640.1 | - | 1 - 2 | 883 | 63 | ||
| BC165435.1 | - | 1 - 2 | 228 | 63 | ||
More Information
Set Membership: |
|

fgf12a
Genomic Map