Assay Details
Gene Symbol: thraa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131396.1 | NP_571471.1 | 3 - 4 | 481 | 90 | |
XM_005163985.4 | XP_005164042.1 | 2 - 3 | 209 | 90 | ||
XM_005163986.4 | XP_005164043.1 | 3 - 4 | 1086 | 90 | ||
XM_009299516.3 | XP_009297791.1 | 2 - 3 | 209 | 90 | ||
XM_021469328.1 | XP_021325003.1 | 3 - 4 | 304 | 90 | ||
GenBank mRNA | BC096778.1 | - | 3 - 4 | 481 | 90 | |
BC164729.1 | - | 2 - 3 | 150 | 90 | ||
U54796.1 | - | 3 - 4 | 481 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
