Assay Details
| Gene Symbol: vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213398.1 | NP_998563.1 | 3 - 4 | 319 | 123 | |
| XM_005159424.4 | XP_005159481.1 | 4 - 5 | 645 | 123 | ||
| GenBank mRNA | AY851290.1 | - | 4 - 5 | 501 | 123 | |
| BC059687.1 | - | 3 - 4 | 319 | 123 | ||
More Information
Set Membership: |
|

vamp1
Genomic Map