Assay Details
| Gene Symbol: rps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012316.1 | NP_001012316.1 | 2 - 3 | 105 | 69 | |
| NM_201216.1 | NP_957510.1 | 83 | 69 | |||
| XM_005156806.4 | XP_005156863.1 | 2 - 3 | 183 | 69 | ||
| XM_005156807.4 | XP_005156864.1 | 2 - 3 | 183 | 69 | ||
| XM_021480547.1 | XP_021336222.1 | 2 - 3 | 311 | 69 | ||
| GenBank mRNA | AY423012.1 | - | 83 | 69 | ||
| BC081494.1 | - | 2 - 3 | 105 | 69 | ||
| BC094973.1 | - | 2 - 3 | 105 | 69 | ||
| BC165902.1 | - | 1 - 2 | 106 | 69 | ||
More Information
Set Membership: |
|

rps24
Genomic Map