Assay Details
| Gene Symbol: kbtbd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033917.2 | NP_001029089.1 | 3 - 4 | 1511 | 60 | |
| GenBank mRNA | BC103489.1 | - | 3 - 4 | 1515 | 60 | |
More Information
Set Membership: |
|

kbtbd12
Genomic Map