Assay Details
| Gene Symbol: myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309488.1 | NP_001296417.1 | 4 - 5 | 667 | 62 | |
| GenBank mRNA | BC114295.1 | - | 4 - 5 | 573 | 62 | |
More Information
Set Membership: |
|

myg1
Genomic Map