Assay Details
| Gene Symbol: ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077451.2 | NP_001070919.2 | 6 - 7 | 729 | 100 | |
| XM_005169311.4 | XP_005169368.1 | 6 - 7 | 808 | 100 | ||
| GenBank mRNA | BC124358.1 | - | 6 - 7 | 728 | 100 | |
More Information
Set Membership: |
|

ccdc120
Genomic Map