Assay Details
| Gene Symbol: srpk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199650.1 | NP_955944.1 | 9 - 10 | 819 | 97 | |
| XM_017357280.2 | XP_017212769.2 | 9 - 10 | 860 | 97 | ||
| GenBank mRNA | BC056825.1 | - | 9 - 10 | 819 | 97 | |
More Information
Set Membership: |
|

srpk1a
Genomic Map